Anti-CD59 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590331EA 568 EUR
ORIGTA590331
Anti-CD59 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot 1:5000-20000, ELISA 1:100-1:2000

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR