Anti-Rapgef2 Polyclonal Antibody

Leverancier: OriGene
ORIGTA329531EA 500 EUR
ORIGTA329531
Anti-Rapgef2 Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Rapgef2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR