Anti-RAB22A Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590265EA 660 EUR
ORIGTA590265
Anti-RAB22A Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot 1:5000-20000, ELISA 1:100-1:2000

Type: Primary
Antigen: RAB22A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Dog, Rat, Monkey, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR