Anti-COL8A1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590478EA 548 EUR
ORIGTA590478
Anti-COL8A1 Rabbit Polyclonal Antibody
Recommended Dilutions: ELISA: 1:100-1:2000; IF/ICC 1:250-1:2000

Type: Primary
Antigen: COL8A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR