Anti-ALDH4A1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590378EA 684 EUR
ORIGTA590378
Anti-ALDH4A1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot 1:20000, IHC 1:150, ELISA 1:100-1:2000

Type: Primary
Antigen: ALDH4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR