Anti-ZNF883 Polyclonal Antibody

Leverancier: OriGene
ORIGTA329369EA 518 EUR
ORIGTA329369
Anti-ZNF883 Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ZNF883
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR