Anti-CBX4 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA329637EA 500 EUR
ORIGTA329637
Anti-CBX4 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR