Anti-MUC16 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590450EA 603 EUR
ORIGTA590450
Anti-MUC16 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000; IHC: 1:10-1:2000; IHC-P 1:250-1:2000

Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR