Anti-Ano1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA328784EA 952 EUR
ORIGTA328784
Anti-Ano1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: Ano1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR