Anti-Slc18a3 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA328797EA 981 EUR
ORIGTA328797
Anti-Slc18a3 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: Slc18a3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR