Anti-ATF6 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA329384EA 575 EUR
ORIGTA329384
Anti-ATF6 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ATF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR