Anti-ANPEP Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA329672EA 500 EUR
ORIGTA329672
Anti-ANPEP Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ANPEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Goat
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR