Anti-EEA1 Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA329677EA 500 EUR
ORIGTA329677
Anti-EEA1 Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: EEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR