Anti-VPS16 Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA329683EA 502 EUR
ORIGTA329683
Anti-VPS16 Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: VPS16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, , Mouse, Mouse, Pig, Rat,
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR