Anti-TUBB2A Mouse Monoclonal Antibody [clone: 3B7]

Leverancier: OriGene
ORIGTA506588EA 562 EUR
ORIGTA506588
Anti-TUBB2A Mouse Monoclonal Antibody [clone: 3B7]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: TUBB2A
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR