Anti-MMP13 Mouse Monoclonal Antibody [clone: 1F1]

Leverancier: OriGene
ORIGTA506692EA 562 EUR
ORIGTA506692
Anti-MMP13 Mouse Monoclonal Antibody [clone: 1F1]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR