Anti-VCP Goat Polyclonal Antibody

Leverancier: OriGene
ORIGTA329667EA 547 EUR
ORIGTA329667
Anti-VCP Goat Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: VCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, , Mouse, Rat, saccharomyces cerevisiae
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR