Anti-PDZD2 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590362EA 625 EUR
ORIGTA590362
Anti-PDZD2 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: ELISA: 1:100-1:2000; IHC: 1:250

Type: Primary
Antigen: PDZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR