Anti-Grik1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA328827EA 953 EUR
ORIGTA328827
Anti-Grik1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:2000; Flow Cytometry: 1:50-1:600

Type: Primary
Antigen: Grik1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR