Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 3G4]

Leverancier: OriGene
ORIGTA507036EA 616 EUR
ORIGTA507036
Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 3G4]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: SERPINB3
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR