Anti-APOA5 Mouse Monoclonal Antibody [clone: 1B4]

Leverancier: OriGene
ORIGTA507096EA 562 EUR
ORIGTA507096
Anti-APOA5 Mouse Monoclonal Antibody [clone: 1B4]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: APOA5
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR