Anti-FGG Mouse Monoclonal Antibody [clone: 2D2]

Leverancier: OriGene
ORIGTA507132EA 616 EUR
ORIGTA507132
Anti-FGG Mouse Monoclonal Antibody [clone: 2D2]
Antilichamen
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150

Type: Primary
Antigen: FGG
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR