Anti-SON Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA590409EA 625 EUR
ORIGTA590409
Anti-SON Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000

Type: Primary
Antigen: SON
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR