Anti-ATAT1 Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA329480EA 500 EUR
ORIGTA329480
Anti-ATAT1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ATAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR