Anti-ICAM3 Mouse Monoclonal Antibody [clone: 4G8]

Leverancier: OriGene
ORIGTA507167EA 616 EUR
ORIGTA507167
Anti-ICAM3 Mouse Monoclonal Antibody [clone: 4G8]
Antilichamen
Recommended Dilutions: Western Blot: 1:400-4000

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR