Anti-SHBG Mouse Monoclonal Antibody [clone: 1H10]

Leverancier: OriGene
ORIGTA507187EA 562 EUR
ORIGTA507187
Anti-SHBG Mouse Monoclonal Antibody [clone: 1H10]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR