Anti-SCARB1 Mouse Monoclonal Antibody [clone: 1E1]

Leverancier: OriGene
ORIGTA507188EA 562 EUR
ORIGTA507188
Anti-SCARB1 Mouse Monoclonal Antibody [clone: 1E1]
Antilichamen
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: SCARB1
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR