Anti-VEGFB Mouse Monoclonal Antibody [clone: 1H9]

Leverancier: OriGene
ORIGTA507205EA 616 EUR
ORIGTA507205
Anti-VEGFB Mouse Monoclonal Antibody [clone: 1H9]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: VEGFB
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR