Anti-SMAD1 Mouse Monoclonal Antibody [clone: 1C2]

Leverancier: OriGene
ORIGTA507243EA 562 EUR
ORIGTA507243
Anti-SMAD1 Mouse Monoclonal Antibody [clone: 1C2]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: SMAD1
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR