Anti-Syp Rabbit Polyclonal Antibody

Leverancier: OriGene
ORIGTA328896EA 952 EUR
ORIGTA328896
Anti-Syp Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:100-1:3000

Type: Primary
Antigen: Syp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR