Anti-ADD1 Rabbit Polyclonal Antibody [clone: Poly6170]

Leverancier: OriGene
ORIGTA328057EA 466 EUR
ORIGTA328057
Anti-ADD1 Rabbit Polyclonal Antibody [clone: Poly6170]
Type: Primary
Antigen: ADD1
Clonality: Polyclonal
Clone: Poly6170
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR