Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]

Leverancier: OriGene
ORIGTA328062EA 486 EUR
ORIGTA328062
Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]
Antilichamen
Type: Primary
Antigen: PRC1
Clonality: Monoclonal
Clone: 6G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR