Anti-NACC1 Rabbit Polyclonal Antibody [clone: Poly6404]

Leverancier: OriGene
ORIGTA328089EA 466 EUR
ORIGTA328089
Anti-NACC1 Rabbit Polyclonal Antibody [clone: Poly6404]
Type: Primary
Antigen: NACC1
Clonality: Polyclonal
Clone: Poly6404
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR