Anti-SCP2 Mouse Monoclonal Antibody [clone: 1E4]

Leverancier: OriGene
ORIGTA507247EA 562 EUR
ORIGTA507247
Anti-SCP2 Mouse Monoclonal Antibody [clone: 1E4]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: SCP2
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR