Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2F6]

Leverancier: OriGene
ORIGTA507280EA 562 EUR
ORIGTA507280
Anti-TXNDC5 Mouse Monoclonal Antibody [clone: 2F6]
Antilichamen
Recommended Dilutions: Western Blot: 1:400-4000

Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR