Anti-HSCB Mouse Monoclonal Antibody [clone: 2B7]

Leverancier: OriGene
ORIGTA507285EA 562 EUR
ORIGTA507285
Anti-HSCB Mouse Monoclonal Antibody [clone: 2B7]
Antilichamen
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: HSCB
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR