Anti-WDHD1 Mouse Monoclonal Antibody [clone: 20G10]

Leverancier: OriGene
ORIGTA328108EA 514 EUR
ORIGTA328108
Anti-WDHD1 Mouse Monoclonal Antibody [clone: 20G10]
Antilichamen
Type: Primary
Antigen: WDHD1
Clonality: Monoclonal
Clone: 20G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR