Anti-IgG2b Rabbit Polyclonal Antibody (DyLight® 488)

Leverancier: Rockland Immunochemicals
ROCK610-441-042EA 196 EUR
ROCK610-441-042
Anti-IgG2b Rabbit Polyclonal Antibody (DyLight® 488)
Antilichamen
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR