Mononucleosis test
Leverancier: SureScreen Diagnostics
SUREMONOCEA 49.5 EUR
SUREMONOC
Mononucleosis test
Mononucleosis, or glandular fever, is a disease caused by the Epstein-Barr virus, which is a member of the herpes family. The virus can remain dormant but become active when the individual is fatigued or in poor health.
This test is ideal for testing a patient who is exhibiting symptoms such as a sore throat, swollen glands and experiencing fatigue to find out if they are suffering from mononucleosis related illness.