Tissue DNA Mini Kits, peqGOLD
PEQLEMR503005 PEQL12-3496-00 PEQLEMR503200 PEQL12-3496-02 PEQL12-3496-01 PEQLEMR503050
Tissue DNA Mini Kits, peqGOLD
Tissue DNA Mini kits for the quick and reliable isolation of high quality DNA from a wide range of animal and human cells and tissue. An optimised MicroSpin option is available for very small sample amounts.
  • No need for organic extractions
  • No need for time-consuming alcohol precipitations
  • Total enzymatic degradation of cellular RNA
  • Efficient separation of enzyme inhibitors and contaminants
  • Final elution with low salt buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids
Leveringsinformatie: Kits contain either PerfectBind DNA columns (Safety or Classic Line*) or PerfectBind MicroSpin DNA columns. All kits contain collection tubes, DNA Lysis Buffer T, DNA Binding Buffer, DNA Wash Buffer, TE Buffer, Elution Buffer and Proteinase K.