Polydimethylsiloxane emulsion, Antifoam B®, Laboratory Reagent, J.T. Baker®
Leverancier: Avantor
Synoniemen: Siliconenolie
Antifoam B™
B531-07EA 216.72 EUR
B531-01 B531-07 B531-05
Polydimethylsiloxane emulsion, Antifoam B®, Laboratory Reagent, J.T. Baker®
Polydimethylsiloxane
White liquid. Silicone emulsion.
MDL-nummer: MFCD00084411
CAS-nummer: 63148-62-9
Bestel nu
Specificaties testresultaten
For Laboratory,Research,or Manufacturing Use
Nonvolatile Content, For Information Only
Appearance Passes Test