ALFA40802

40802.18EA 578 EUR
40802.18 40802.09
ALFA40802
Density: 2.1
Sensitivity: Hygroscopic
Appearance: White to pale brown
Melting Point: 200°
Applications: Super acid catalyst
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR