Roxithromycin 97%

Leverancier: Thermo Scientific
Waarschuwing

J66213.09EA 145 EUR
J66213.09
Roxithromycin 97%
Roxithromycin

Roxithromycin is used to block bacterial protein synthesis


  • White powder


Clinically, used to treat respiratory tract, urinary and soft tissue infections.


Soluble in water to 18µg/ml

Formule: C₄₁H₇₆N₂O₁₅
Smeltpunt: 111…118 °C
Vlampunt: >110 °C (230 °F)
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00214389
CAS-nummer: 80214-83-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR