Kanamycin disulphate 700 µg/mg
Leverancier: Thermo Scientific
|
Gevaar
|
J66856.06EA
97.6
EUR
J66856.06
Kanamycin disulphate 700 µg/mg
Kanamycin disulphate
Kanamycin sulfate is a broad spectrum minoglycoside-antibiotic.
- Moisture and light sensitive
Soluble in aqueous solutions
Formule:
C₁₈H₃₆N₄O₁₁·2H₂SO₄ MW: 680,65 g/mol Smeltpunt: >260 °C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00143916 CAS-nummer: 64013-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...