1-Chloro-2-butene (mixture of cis and trans isomers) 70% tech.

Leverancier: Thermo Scientific
Gevaar

ACRO154801000EA 61.9 EUR
ACRO154801000 ACRO154805000
1-Chloro-2-butene (mixture of cis and trans isomers) 70% tech.
1-Chloro-2-butene (mixture of cis and trans isomers)
Formule: C₄H₇Cl
MW: 90,55 g/mol
Kookpunt: 83…84 °C (1013 hPa)
Smeltpunt: –65 °C
Dichtheid: 0,92 g/cm³
Vlampunt: –15 °C
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00000987
CAS-nummer: 591-97-9
UN: 2924
ADR: 3,II

Bestel nu

Specificaties testresultaten

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4340 (20°C, 589 nm)
GC ≥65 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR