Tributylphosphine 95%, AcroSeal™

Leverancier: Thermo Scientific
Gevaar

427301000.EA 54.5 EUR
427301000.
Tributylphosphine 95%, AcroSeal™
Tributylphosphine
Formule: C₁₂H₂₇P
MW: 202,32 g/mol
Kookpunt: 240 °C (1013 hPa)
Smeltpunt: –65 °C
Dichtheid: 0,81 g/cm³
Vlampunt: 37 °C
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00009462
CAS-nummer: 998-40-3
UN: 3254
ADR: 4.2,I

Bestel nu

Specificaties testresultaten

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR