2',6'-Difluoroacetophenone 98%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO214290050EA
101
EUR
ACRO214290050
2',6'-Difluoroacetophenone 98%
2',6'-Difluoroacetophenone
Formule:
C₈H₆F₂O MW: 156,13 g/mol Kookpunt: 76…79 °C (15 mmHg) Dichtheid: 1,19 g/cm³ Vlampunt: 76 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00000328 CAS-nummer: 13670-99-0 |
Specificaties testresultaten
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...