Dipentene, fraction of terpene hydrocarbons fraction of terpene hydrocarbons, pure
|
Gevaar
|
ACRO390310025EA
60.5
EUR
ACRO390310025
ACRO390310010
Dipentene, fraction of terpene hydrocarbons fraction of terpene hydrocarbons, pure
Dipentene, fraction of terpene hydrocarbons
Formule:
C₁₀H₁₆ Kookpunt: 174 °C (760 mmHg) Dichtheid: 0,868 g/cm³ Vlampunt: 42 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00062992 CAS-nummer: 68956-56-9 UN: 2052 ADR: 3,III |
Specificaties testresultaten
Appearance | Clear liquid |
Refractive index | 1.4730 to 1.4780 (20°C, 589 nm) |
Density | (20°C) 0.855 to 0.870 kg/l |
Color scale | ≤50 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...