3,3-Dimethoxybutan-2-one 98%
Leverancier: Thermo Scientific
|
Waarschuwing
|
ACRO399720250EA
275
EUR
ACRO399720250
3,3-Dimethoxybutan-2-one 98%
3,3-Dimethoxybutan-2-one
Formule:
C₆H₁₂O₃ MW: 132,159 g/mol Kookpunt: 145…146 °C (760 mmHg) Dichtheid: 0,987 g/cm³ Vlampunt: 45 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00075266 CAS-nummer: 21983-72-2 UN: 1224 ADR: 3,III |
Specificaties testresultaten
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4060 to 1.4080 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...