Sample collection cards, Ahlstrom-Munksjö GenCollect™ and GenCollect™ 2.0
BINZ8.560.0000.B-NEA 0 EUR
BINZ8.560.0000.B-N BINZ8.562.0002.B-N BINZ8.562.0001.B-N BINZ8.563.0000.B-N BINZ8.560.0001.B-N BINZ8.560.0002.B-N BINZ8.561.0001.B-N BINZ8.563.0002.B-N BINZ8.561.0000.B-N BINZ8.562.0000.B-N BINZ8.563.0001.B-N BINZ8.561.0002.B-N
Sample collection cards, Ahlstrom-Munksjö GenCollect™ and GenCollect™ 2.0
GenCollect™ and GenCollect™ 2.0 cards, made of high purity cellulose, provide a rapid and safe way to collect, transport and store biological samples at ambient temperatures. Nucleic acids can be easily extracted from the biosamples for subsequent analyses.
  • Simple collection
  • Nucleic acids stablisation for short term ambient storage
  • Cost and time-saving DNA purification
  • Faster downstream analyses
  • Ready to use biosample collection card
GenCollect™ card is a chemical free card designed for efficient extraction and purification of DNA. GenCollect™ 2.0 card is a chemically treated card that lyses cells and preserves biosamples from microbiological contamination. It is fully compatible for direct DNA amplification, eliminating the extraction step.
The next generation GenCollect™ 2.0 and GenCollect™ Colour 2.0 cards offer additional features in order to allow direct amplification of DNA from a paper disc and to prevent the growth of microorganisms during the short-term storage at ambient temperature.
Bestel nu